Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
prx-6
Entrez Gene ID
266912
Gene Name
AAA domain-containing protein
Gene Aliases
CELE_F39G3.7
Chromosome Location
Chr.V: 4715388 - 4720148 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071867.5 NP_504268.1 4 - 5 1121 156
GenBank mRNA AB010968.1 - 4 - 5 1085 156

View Full Product Details