Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
tig-2
Entrez Gene ID
178864
Gene Name
TGF_BETA_2 domain-containing protein
Gene Aliases
CELE_F39G3.8
Chromosome Location
Chr.V: 4726721 - 4730234 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071870.5 NP_504271.1 6 - 7 807 63

View Full Product Details