Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cwp-3
Entrez Gene ID
183296
Gene Name
Coexpressed With Polycystins
Gene Aliases
CELE_C37H5.4
Chromosome Location
Chr.V: 4842836 - 4843845 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071895.2 NP_504296.2 1 - 2 64 72

View Full Product Details