Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ncx-9
Entrez Gene ID
182576
Gene Name
Mitochondrial sodium/calcium exchanger protein
Gene Aliases
CELE_C13D9.8
Chromosome Location
Chr.V: 4991342 - 4995255 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071941.3 NP_504342.2 7 - 8 1405 124

View Full Product Details