Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpt-4
Entrez Gene ID
178889
Gene Name
Carn_acyltransf domain-containing protein
Gene Aliases
CELE_K11D12.4
Chromosome Location
Chr.V: 5023706 - 5027723 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071947.5 NP_504348.1 2 - 3 227 82

View Full Product Details