Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cwp-4
Entrez Gene ID
178894
Gene Name
Coexpressed With Polycystins
Gene Aliases
CELE_K11D12.1
Chromosome Location
Chr.V: 5053619 - 5056474 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_071955.6 NP_504356.2 5 - 6 983 73

View Full Product Details