Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-68
Entrez Gene ID
191141
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_ZC317.5
Chromosome Location
Chr.V: 5463791 - 5465435 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072043.1 NP_504444.1 4 - 5 831 172

View Full Product Details