Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-47
Entrez Gene ID
178933
Gene Name
Nuclear hormone receptor family member nhr-47
Gene Aliases
CELE_C24G6.4
Chromosome Location
Chr.V: 5505253 - 5509956 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072054.6 NP_504455.1 6 - 7 893 102
GenBank mRNA AY204170.1 - 6 - 7 760 102
U41296.1 - 6 - 7 791 102

View Full Product Details