Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grl-1
Entrez Gene ID
182850
Gene Name
Ground-like domain-containing protein
Gene Aliases
CELE_C24G6.7
Chromosome Location
Chr.V: 5530725 - 5533855 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072059.2 NP_504460.1 3 - 4 758 72

View Full Product Details