Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rga-3
Entrez Gene ID
178961
Gene Name
Rho-GAP domain-containing protein
Gene Aliases
CELE_K09H11.3
Chromosome Location
Chr.V: 5717885 - 5722089 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072102.4 NP_504503.1 1 - 2 144 83

View Full Product Details