Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
grd-9
Entrez Gene ID
191663
Gene Name
Ground-like domain-containing protein
Gene Aliases
CELE_C04E6.6
Chromosome Location
Chr.V: 5895321 - 5898602 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072134.6 NP_504535.2 7 - 8 1396 149

View Full Product Details