The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_072155.6 | NP_504556.3 | 2 - 3 | 226 | 73 |
GenBank mRNA | DQ340626.1 | - | 1 - 2 | 154 | 73 |