Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpg-4
Entrez Gene ID
178986
Gene Name
Chondroitin proteoglycan 4
Gene Aliases
CELE_C10F3.1
Chromosome Location
Chr.V: 5995844 - 5999353 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072155.6 NP_504556.3 5 - 6 1019 74
GenBank mRNA DQ340626.1 - 4 - 5 947 74

View Full Product Details