Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srh-28
Entrez Gene ID
191850
Gene Name
Serpentine Receptor, class H
Gene Aliases
CELE_ZC404.5
Chromosome Location
Chr.V: 6774261 - 6775808 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380678.2 NP_001367940.1 2 - 3 959 115

View Full Product Details