Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
odr-2
Entrez Gene ID
179098
Gene Name
protein-coding
Gene Aliases
CELE_T01C4.2
Chromosome Location
Chr.V: 7103543 - 7114439 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028918.4 NP_001024089.1 3 - 4 764 144
NM_001028919.5 NP_001024090.1 3 - 4 411 144
NM_001028920.4 NP_001024091.1 4 - 5 451 144
GenBank mRNA AF324050.1 - 4 - 5 440 144
AF324051.1 - 3 - 4 375 144
AF324052.1 - 3 - 4 532 144

View Full Product Details