Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-33
Entrez Gene ID
187943
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T01C4.3
Chromosome Location
Chr.V: 7117452 - 7118896 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322563.1 NP_001309513.1 1 - 2 197 130

View Full Product Details