Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srj-32
Entrez Gene ID
191944
Gene Name
protein-coding
Gene Aliases
CELE_F07G11.5
Chromosome Location
Chr.V: 7296149 - 7297363 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269154.3 NP_001256083.1 1 - 2 257 187
NM_001269155.2 NP_001256084.1 1 - 2 255 187

View Full Product Details