Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-92
Entrez Gene ID
184316
Gene Name
protein-coding
Gene Aliases
CELE_F10G2.6
Chromosome Location
Chr.V: 7324038 - 7325385 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028612.1 NP_001023783.1 2 - 3 220 101

View Full Product Details