Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-9
Entrez Gene ID
187859
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_R13D7.6
Chromosome Location
Chr.V: 7393550 - 7395459 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072492.1 NP_504893.1 1 - 2 211 164

View Full Product Details