Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-68
Entrez Gene ID
184640
Gene Name
protein-coding
Gene Aliases
CELE_F18E3.4
Chromosome Location
Chr.V: 7425259 - 7426694 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028646.1 NP_001023817.1 3 - 4 688 120
NM_001028647.2 NP_001023818.1 3 - 4 688 120

View Full Product Details