Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-69
Entrez Gene ID
184641
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F18E3.5
Chromosome Location
Chr.V: 7427326 - 7428980 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072504.1 NP_504905.1 6 - 7 1090 82

View Full Product Details