Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hint-3
Entrez Gene ID
182943
Gene Name
protein-coding
Gene Aliases
CELE_C26F1.7
Chromosome Location
Chr.V: 7760474 - 7761633 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269160.3 NP_001256089.1 2 - 3 174 95
NM_001269161.2 NP_001256090.1 1 - 2 85 95

View Full Product Details