Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-53
Entrez Gene ID
190824
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_Y97E10B.10
Chromosome Location
Chr.V: 7938605 - 7940399 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072641.4 NP_505042.3 3 - 4 725 164

View Full Product Details