Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oma-2
Entrez Gene ID
179183
Gene Name
CCCH-type zinc finger protein oma-2
Gene Aliases
CELE_ZC513.6
Chromosome Location
Chr.V: 8029437 - 8031214 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072668.8 NP_505069.1 4 - 5 637 62

View Full Product Details