Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-44
Entrez Gene ID
179276
Gene Name
Nuclear hormone receptor family member nhr-44
Gene Aliases
CELE_T19A5.4
Chromosome Location
Chr.V: 8961882 - 8966185 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072912.7 NP_505313.1 11 - 12 1748 144

View Full Product Details