Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-85
Entrez Gene ID
191992
Gene Name
Seven TM Receptor
Gene Aliases
CELE_F59E11.15
Chromosome Location
Chr.V: 8984525 - 8985840 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072920.1 NP_505321.1 3 - 4 488 126

View Full Product Details