Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-2
Entrez Gene ID
187548
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_R03H4.2
Chromosome Location
Chr.V: 9009475 - 9010830 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072929.2 NP_505330.1 3 - 4 599 175

View Full Product Details