Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-41
Entrez Gene ID
187551
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_R03H4.5
Chromosome Location
Chr.V: 9019305 - 9022178 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072933.2 NP_505334.1 4 - 5 622 154

View Full Product Details