Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ptd-2
Entrez Gene ID
179307
Gene Name
SSD domain-containing protein
Gene Aliases
CELE_F07C3.1
Chromosome Location
Chr.V: 9250232 - 9254328 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_072995.3 NP_505396.2 12 - 13 1848 114

View Full Product Details