Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
coq-6
Entrez Gene ID
179316
Gene Name
Ubiquinone biosynthesis monooxygenase COQ6, mitochondrial
Gene Aliases
CELE_K07B1.2
Chromosome Location
Chr.V: 9336762 - 9338858 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073014.5 NP_505415.2 1 - 2 110 70

View Full Product Details