Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpa-3
Entrez Gene ID
179319
Gene Name
Guanine nucleotide-binding protein alpha-3 subunit
Gene Aliases
CELE_E02C12.5
Chromosome Location
Chr.V: 9356579 - 9359774 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322582.2 NP_001309498.1 3 - 4 619 142
NR_136369.1 3 - 4 595 142

View Full Product Details