Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
atl-1
Entrez Gene ID
179352
Gene Name
protein-coding
Gene Aliases
CELE_T06E4.3
Chromosome Location
Chr.V: 9634861 - 9645595 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073086.5 NP_505487.3 9 - 10 3465 79

View Full Product Details