Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-40
Entrez Gene ID
187650
Gene Name
protein-coding
Gene Aliases
CELE_R07B5.3
Chromosome Location
Chr.V: 9832646 - 9834000 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269210.2 NP_001256139.1 1 - 2 430 83

View Full Product Details