Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-12
Entrez Gene ID
179403
Gene Name
protein-coding
Gene Aliases
CELE_R04B5.4
Chromosome Location
Chr.V: 10082124 - 10084049 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001129511.5 NP_001122983.1 3 - 4 899 126
NM_001276926.3 NP_001263855.1 3 - 4 899 126
NM_073188.6 NP_505589.1 3 - 4 893 126

View Full Product Details