Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-28
Entrez Gene ID
183705
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C51E3.3
Chromosome Location
Chr.V: 10152578 - 10154832 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322584.2 NP_001309452.1 3 - 4 340 101

View Full Product Details