Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
che-12
Entrez Gene ID
179433
Gene Name
protein-coding
Gene Aliases
CELE_B0024.8
Chromosome Location
Chr.V: 10299472 - 10307383 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269275.3 NP_001256204.1 4 - 5 539 93

View Full Product Details