Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-33
Entrez Gene ID
179464
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_C35A5.2
Chromosome Location
Chr.V: 10493407 - 10496477 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073291.9 NP_505692.2 5 - 6 1079 110

View Full Product Details