Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ocr-1
Entrez Gene ID
185101
Gene Name
protein-coding
Gene Aliases
CELE_F28H7.10
Chromosome Location
Chr.V: 10764366 - 10768590 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001348583.2 NP_001335506.1 12 - 13 1880 150

View Full Product Details