Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srsx-33
Entrez Gene ID
188584
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_T19B10.10
Chromosome Location
Chr.V: 11251415 - 11252529 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073457.2 NP_505858.2 2 - 3 215 72

View Full Product Details