Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cnb-1
Entrez Gene ID
179572
Gene Name
protein-coding
Gene Aliases
CELE_F55C10.1
Chromosome Location
Chr.V: 11375742 - 11378006 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269389.2 NP_001256318.1 5 - 6 510 82
GenBank mRNA AY190131.1 - 4 - 5 461 82

View Full Product Details