Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-16
Entrez Gene ID
186922
Gene Name
Zinc metalloproteinase nas-16
Gene Aliases
CELE_K03B8.1
Chromosome Location
Chr.V: 11392320 - 11393508 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073488.2 NP_505889.2 1 - 2 163 141

View Full Product Details