Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-85
Entrez Gene ID
185457
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F38B7.4
Chromosome Location
Chr.V: 11552679 - 11554223 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073529.3 NP_505930.2 4 - 5 646 80

View Full Product Details