Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rhr-2
Entrez Gene ID
179606
Gene Name
Ammonium_transp domain-containing protein
Gene Aliases
CELE_B0240.1
Chromosome Location
Chr.V: 11727867 - 11730160 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073560.6 NP_505961.1 3 - 4 340 99

View Full Product Details