Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
npp-22
Entrez Gene ID
179607
Gene Name
Nucleoporin ndc-1
Gene Aliases
CELE_B0240.4
Chromosome Location
Chr.V: 11731113 - 11733472 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073561.7 NP_505962.2 5 - 6 925 125

View Full Product Details