Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-76
Entrez Gene ID
187336
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_K12G11.5
Chromosome Location
Chr.V: 11891812 - 11893451 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073592.2 NP_505993.2 2 - 3 238 112

View Full Product Details