Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
kup-1
Entrez Gene ID
179651
Gene Name
Protein kup-1
Gene Aliases
CELE_F10C2.2
Chromosome Location
Chr.V: 12014433 - 12040873 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383418.1 1 - 2 101 69
GenBank mRNA L12247.1 - 1 - 2 123 69

View Full Product Details