Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elpc-3
Entrez Gene ID
179669
Gene Name
Elongator complex protein 3
Gene Aliases
CELE_ZK863.3
Chromosome Location
Chr.V: 12179497 - 12181410 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073654.5 NP_506055.2 2 - 3 479 76

View Full Product Details