Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
myo-3
Entrez Gene ID
179676
Gene Name
Myosin-3
Gene Aliases
CELE_K12F2.1
Chromosome Location
Chr.V: 12226816 - 12234343 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073664.6 NP_506065.2 2 - 3 396 139

View Full Product Details