Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
twk-24
Entrez Gene ID
179682
Gene Name
TWiK family of potassium channels
Gene Aliases
CELE_F55C5.3
Chromosome Location
Chr.V: 12265707 - 12269325 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380782.1 NP_001367093.1 2 - 3 429 158

View Full Product Details