Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fkb-4
Entrez Gene ID
179752
Gene Name
Peptidylprolyl isomerase
Gene Aliases
CELE_ZC455.10
Chromosome Location
Chr.V: 12785153 - 12787126 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073796.6 NP_506197.1 3 - 4 724 157

View Full Product Details